siRNA Details
| Gene Symbol: PPM1N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080401.1 | NP_001073870.1 | 3, 4 | 1173 | ||
| GenBank mRNA | AK097444.1 | - | 2, 3 | 1576 | ||
| BC062452.1 | - | 3, 4 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPM1N| Gene Symbol: PPM1N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080401.1 | NP_001073870.1 | 3, 4 | 1173 | ||
| GenBank mRNA | AK097444.1 | - | 2, 3 | 1576 | ||
| BC062452.1 | - | 3, 4 | 351 | |||