siRNA Details
| Gene Symbol: ZNF784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203374.1 | NP_976308.1 | 2 | 787 | ||
| GenBank mRNA | AK074859.1 | - | 2 | 778 | ||
| AK309967.1 | - | 2 | 815 | |||
| BC065822.1 | - | 2 | 787 | 6185645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF784| Gene Symbol: ZNF784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203374.1 | NP_976308.1 | 2 | 787 | ||
| GenBank mRNA | AK074859.1 | - | 2 | 778 | ||
| AK309967.1 | - | 2 | 815 | |||
| BC065822.1 | - | 2 | 787 | 6185645 | ||