siRNA Details
| Gene Symbol: ZNF681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138286.2 | NP_612143.2 | 4 | 949 | ||
| GenBank mRNA | AK056088.1 | - | 3 | 747 | ||
| AK122869.1 | - | 4 | 953 | |||
| BC136346.1 | - | 4 | 828 | 9020333 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF681| Gene Symbol: ZNF681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138286.2 | NP_612143.2 | 4 | 949 | ||
| GenBank mRNA | AK056088.1 | - | 3 | 747 | ||
| AK122869.1 | - | 4 | 953 | |||
| BC136346.1 | - | 4 | 828 | 9020333 | ||