siRNA Details
| Gene Symbol: ZNF555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172775.1 | NP_001166246.1 | 4 | 1859 | ||
| NM_152791.4 | NP_690004.4 | 4 | 1862 | |||
| XM_011527716.2 | XP_011526018.1 | 4 | 1862 | |||
| XM_017026375.1 | XP_016881864.1 | 4 | 1862 | |||
| GenBank mRNA | AK056659.1 | - | 1 | 1160 | ||
| AK093163.1 | - | 6 | 1595 | |||
| AK292954.1 | - | 4 | 1807 | |||
| AL832140.1 | - | 4 | 1823 | |||
| BC022022.2 | - | 4 | 1813 | 4822197 | ||
| BX647423.1 | - | 4 | 1820 | |||

ZNF555
Genomic Map