siRNA Details
| Gene Symbol: LELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010857.2 | NP_001010857.1 | 2 | 245 | ||
| GenBank mRNA | BC087854.1 | - | 2 | 185 | 30387074 | |
| BC130507.1 | - | 2 | 225 | 40146538 | ||
| BC130511.1 | - | 2 | 225 | 40146542 | ||
| DB069578.1 | - | 2 | 245 | |||
| DB086762.1 | - | 2 | 245 | |||

LELP1
Genomic Map