siRNA Details
| Gene Symbol: RNF187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010858.2 | NP_001010858.2 | 3 | 693 | ||
| GenBank mRNA | BC012758.2 | - | 3 | 667 | ||
| BC064481.1 | - | 3 | 227 | |||
| BC080645.1 | - | 3 | 227 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF187| Gene Symbol: RNF187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010858.2 | NP_001010858.2 | 3 | 693 | ||
| GenBank mRNA | BC012758.2 | - | 3 | 667 | ||
| BC064481.1 | - | 3 | 227 | |||
| BC080645.1 | - | 3 | 227 | |||