siRNA Details
| Gene Symbol: CDPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207327.4 | NP_997210.3 | 2 | 81 | ||
| XM_011529960.1 | XP_011528262.1 | 2 | 464 | |||
| XM_011529962.2 | XP_011528264.1 | 2 | 72 | |||
| XM_011529963.1 | XP_011528265.1 | 2 | 464 | |||
| XM_011529964.1 | XP_011528266.1 | 2 | 464 | |||
| XM_011529965.2 | XP_011528267.1 | 2 | 397 | |||
| GenBank mRNA | BC036462.1 | - | 2 | 79 | ||
| BC045796.2 | - | 11 | ||||
| BC067871.1 | - | 2 | 81 | 30338205 | ||
| BX648331.1 | - | 2 | 89 | |||

CDPF1
Genomic Map