siRNA Details
| Gene Symbol: CTSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317331.1 | NP_001304260.1 | 3, 4 | 420 | ||
| NM_001910.3 | NP_001901.1 | 4, 5 | 577 | |||
| NM_148964.2 | NP_683865.1 | 4, 5 | 577 | |||
| GenBank mRNA | AJ250716.1 | - | 4, 5 | 563 | ||
| AJ250717.1 | - | 4, 5 | 563 | |||
| AK292057.1 | - | 4, 5 | 507 | |||
| AK298069.1 | - | 3, 4 | 273 | |||
| BC042537.1 | - | 4, 5 | 544 | 5174814 | ||
| J05036.1 | - | 4, 5 | 508 | |||

CTSE
Genomic Map