siRNA Details
| Gene Symbol: CPNE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308388.1 | NP_001295317.1 | 19, 20 | 1601 | ||
| NM_153635.2 | NP_705899.2 | 19, 20 | 1582 | |||
| XM_011533386.2 | XP_011531688.1 | 18, 19 | 1758 | |||
| XM_017005747.1 | XP_016861236.1 | 18, 19 | 1342 | |||
| GenBank mRNA | AA758411.1 | - | 2, 3 | 466 | ||
| BC035735.1 | - | 19, 20 | 1582 | 5729888 | ||
| BC130375.1 | - | 19, 20 | 1453 | 40146406 | ||
| BU101942.1 | - | 2, 3 | 205 | |||
| CA307982.1 | - | 2, 3 | 481 | |||

CPNE9
Genomic Map