siRNA Details
| Gene Symbol: C3orf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152533.2 | NP_689746.1 | 4 | 693 | ||
| XM_017005751.1 | XP_016861240.1 | 4 | 663 | |||
| XM_017005752.1 | XP_016861241.1 | 4 | 525 | |||
| GenBank mRNA | BC032025.1 | - | 4 | 643 | 5165128 | |

C3orf22
Genomic Map