siRNA Details
| Gene Symbol: CX3CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171171.1 | NP_001164642.1 | 2 | 1250 | ||
| NM_001171172.1 | NP_001164643.1 | 2 | 1210 | |||
| NM_001171174.1 | NP_001164645.1 | 2 | 1146 | |||
| NM_001337.3 | NP_001328.1 | 2 | 1102 | |||
| GenBank mRNA | AK225764.1 | - | 2 | 1103 | ||
| AK307911.1 | - | 3 | 900 | |||
| AK312373.1 | - | 2 | 1101 | |||
| BC028078.1 | - | 2 | 1086 | 5216452 | ||
| U20350.1 | - | 2 | 1097 | |||
| U28934.1 | - | 2 | 1055 | |||

CX3CR1
Genomic Map