siRNA Details
| Gene Symbol: MGAT4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277353.1 | NP_001264282.1 | 1, 2 | 243 | ||
| XM_011531650.2 | XP_011529952.1 | 1, 2 | 1695 | |||
| XM_011531654.2 | XP_011529956.1 | 1, 2 | 1695 | |||
| XM_011531656.2 | XP_011529958.1 | 1, 2 | 1695 | |||
| XM_011531659.1 | XP_011529961.1 | 1, 2 | 1695 | |||
| XM_011531661.1 | XP_011529963.1 | 1, 2 | 1695 | |||
| XM_011531662.1 | XP_011529964.1 | 1, 2 | 1695 | |||
| XM_011531663.2 | XP_011529965.1 | 1, 2 | 1689 | |||
| GenBank mRNA | BI826002.1 | - | 1, 2 | 219 | ||
| DB462368.1 | - | 1, 2 | 244 | |||

MGAT4D
Genomic Map