siRNA Details
| Gene Symbol: JAKMIP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038902.1 | 3 | 336 | |||
| GenBank mRNA | AK027578.1 | - | 3 | 336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

JAKMIP2-AS1| Gene Symbol: JAKMIP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038902.1 | 3 | 336 | |||
| GenBank mRNA | AK027578.1 | - | 3 | 336 | ||