siRNA Details
| Gene Symbol: CCDC167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138493.2 | NP_612502.1 | 1, 2 | 101 | ||
| GenBank mRNA | BC003515.1 | - | 1, 2 | 46 | ||
| BC104835.1 | - | 1, 2 | 97 | 8143838 | ||
| BC108655.1 | - | 1, 2 | 74 | 6053765 | ||
| DA152306.1 | - | 1, 2 | 101 | |||

CCDC167
Genomic Map