siRNA Details
| Gene Symbol: CRYGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144727.2 | NP_653328.1 | 3, 4 | 672 | ||
| NR_131779.1 | 3, 4 | 1024 | ||||
| GenBank mRNA | AF445455.1 | - | 3, 4 | 515 | ||
| BC100878.2 | - | 3, 4 | 510 | 40003204 | ||
| BC100880.2 | - | 3, 4 | 510 | 40003208 | ||
| BC100881.2 | - | 3, 4 | 510 | 40003209 | ||

CRYGN
Genomic Map