siRNA Details
| Gene Symbol: CRYGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308292.1 | NP_001295221.1 | 3 | 792 | ||
| NM_144727.2 | NP_653328.1 | 4 | 781 | |||
| NR_131779.1 | 4 | 1133 | ||||
| GenBank mRNA | AF445455.1 | - | 4 | 624 | ||
| BC043605.1 | - | 4 | 1133 | |||
| BC100878.2 | - | 4 | 619 | 40003204 | ||
| BC100879.3 | - | 3 | 630 | 40003207 | ||
| BC100880.2 | - | 4 | 619 | 40003208 | ||
| BC100881.2 | - | 4 | 619 | 40003209 | ||

CRYGN
Genomic Map