siRNA Details
| Gene Symbol: CYP3A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000765.4 | NP_000756.3 | 2 | 258 | ||
| GenBank mRNA | BC067436.1 | - | 2 | 236 | 7002135 | |
| BX643367.1 | - | 2 | 255 | |||
| D00408.1 | - | 2 | 154 | |||
| Gene Symbol: CYP3A7-CYP3A51P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256497.2 | NP_001243426.2 | 2 | 258 | ||
| GenBank mRNA | AF315325.1 | - | 2 | 151 | ||

CYP3A7
Genomic Map