siRNA Details
Gene Symbol: RBM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053043.2 | NP_444271.2 | 11 | 1907 | ||
XM_005249531.1 | XP_005249588.1 | 12 | 2072 | |||
XM_005249532.1 | XP_005249589.1 | 12 | 2072 | |||
XM_005249533.1 | XP_005249590.1 | 12 | 2042 | |||
XM_005249534.1 | XP_005249591.1 | 11 | 1952 | |||
XM_005249535.1 | XP_005249592.1 | 11 | 1922 | |||
XM_011515840.1 | XP_011514142.1 | 12 | 2072 | |||
GenBank mRNA | AK301179.1 | - | 6 | 854 | ||
AK307515.1 | - | 11 | 1737 | |||
AL137724.1 | - | 10 | 1491 |
