siRNA Details
| Gene Symbol: RBM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053043.2 | NP_444271.2 | 5 | 768 | ||
| XM_005249531.1 | XP_005249588.1 | 5 | 783 | |||
| XM_005249532.1 | XP_005249589.1 | 5 | 783 | |||
| XM_005249533.1 | XP_005249590.1 | 5 | 783 | |||
| XM_005249534.1 | XP_005249591.1 | 5 | 783 | |||
| XM_005249535.1 | XP_005249592.1 | 5 | 783 | |||
| XM_011515840.1 | XP_011514142.1 | 5 | 783 | |||
| GenBank mRNA | AK307515.1 | - | 5 | 598 | ||
| AL137724.1 | - | 3 | 265 | |||
| BC011923.2 | - | 5 | 826 | |||

RBM33
Genomic Map