siRNA Details
| Gene Symbol: AGR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176813.4 | NP_789783.1 | 6 | 395 | ||
| XM_005249627.2 | XP_005249684.1 | 6 | 374 | |||
| XM_011515152.2 | XP_011513454.1 | 6 | 387 | |||
| GenBank mRNA | AY069977.1 | - | 5 | 360 | ||
| AY359065.1 | - | 6 | 379 | |||
| BC058284.1 | - | 6 | 400 | 4694757 | ||
| BM984550.1 | - | 3 | 363 | |||
| BU682778.1 | - | 3 | 369 | |||
| CB242846.1 | - | 3 | 363 | |||

AGR3
Genomic Map