siRNA Details
| Gene Symbol: FAM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103537.1 | 4 | 3037 | |||
| GenBank mRNA | BC037297.2 | - | 4 | 3036 | ||
| BC118644.1 | - | 1 | 853 | |||
| Gene Symbol: FAM87B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103536.1 | 2 | 971 | |||
| GenBank mRNA | AK097327.1 | - | 2 | 971 | ||

FAM87A
Genomic Map