siRNA Details
Gene Symbol: MAMDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206920.2 | NP_996803.2 | 4, 5 | 512 | ||
GenBank mRNA | AY358419.1 | - | 4, 5 | 512 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MAMDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206920.2 | NP_996803.2 | 4, 5 | 512 | ||
GenBank mRNA | AY358419.1 | - | 4, 5 | 512 |