siRNA Details
| Gene Symbol: TTLL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139442.1 | NP_001132914.1 | 3 | 1067 | ||
| NM_194252.2 | NP_919228.2 | 3 | 1067 | |||
| XM_005251729.4 | XP_005251786.1 | 3 | 1445 | |||
| XM_006716966.3 | XP_006717029.1 | 3 | 744 | |||
| XM_017014309.1 | XP_016869798.1 | 3 | 744 | |||
| GenBank mRNA | AF521886.1 | - | 3 | 1067 | ||

TTLL11
Genomic Map