siRNA Details
| Gene Symbol: KIAA1958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287036.1 | NP_001273965.1 | 2 | 253 | ||
| NM_001287038.1 | NP_001273967.1 | 2 | 253 | |||
| NM_133465.3 | NP_597722.1 | 2 | 253 | |||
| XM_011518311.2 | XP_011516613.1 | 2 | 2055 | |||
| GenBank mRNA | AK096613.1 | - | 2 | 254 | ||
| BC105048.1 | - | 2 | 143 | 8144051 | ||
| BC113515.1 | - | 2 | 143 | 8322567 | ||
| BC143424.1 | - | 2 | 143 | 9051932 | ||

KIAA1958
Genomic Map