siRNA Details
| Gene Symbol: FMR1NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152578.2 | NP_689791.1 | 4 | 659 | ||
| GenBank mRNA | AK098602.1 | - | 4 | 644 | ||
| AY211917.1 | - | 4 | 652 | |||
| BC034320.1 | - | 4 | 659 | 4836772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FMR1NB| Gene Symbol: FMR1NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152578.2 | NP_689791.1 | 4 | 659 | ||
| GenBank mRNA | AK098602.1 | - | 4 | 644 | ||
| AY211917.1 | - | 4 | 652 | |||
| BC034320.1 | - | 4 | 659 | 4836772 | ||