siRNA Details
| Gene Symbol: FAM47A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203408.3 | NP_981953.2 | 1 | 1779 | ||
| GenBank mRNA | AK292354.1 | - | 2 | 1724 | ||
| BC026171.2 | - | 2 | 1724 | 4827969 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM47A| Gene Symbol: FAM47A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203408.3 | NP_981953.2 | 1 | 1779 | ||
| GenBank mRNA | AK292354.1 | - | 2 | 1724 | ||
| BC026171.2 | - | 2 | 1724 | 4827969 | ||