siRNA Details
| Gene Symbol: SLC5A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178498.3 | NP_848593.2 | 1 | 582 | ||
| XM_006718156.3 | XP_006718219.1 | 1 | 821 | |||
| XM_011519921.2 | XP_011518223.1 | 1 | 822 | |||
| GenBank mRNA | AY299482.1 | - | 1 | 291 | ||
| BC049207.1 | - | 1 | 554 | 5188979 | ||
| BM924715.1 | - | 1 | 601 | |||

SLC5A12
Genomic Map