siRNA Details
| Gene Symbol: DAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291963.1 | NP_001278892.1 | 3 | 459 | ||
| NM_004394.2 | NP_004385.1 | 4 | 502 | |||
| GenBank mRNA | AK298670.1 | - | 3 | 412 | ||
| AK311783.1 | - | 4 | 381 | |||
| BC002726.2 | - | 4 | 459 | 3631886 | ||
| BI823002.1 | - | 4 | 333 | |||
| BP265421.1 | - | 4 | 463 | |||
| BT006925.1 | - | 3 | 168 | |||
| X76105.1 | - | 4 | 454 | |||

DAP
Genomic Map