siRNA Details
| Gene Symbol: DBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004395.3 | NP_004386.2 | 2 | 296 | ||
| NM_080881.2 | NP_543157.1 | 3 | 412 | |||
| XM_005265827.3 | XP_005265884.1 | 2 | 243 | |||
| XM_006714826.2 | XP_006714889.1 | 2 | 133 | |||
| XM_011534446.2 | XP_011532748.1 | 3 | 415 | |||
| XM_011534447.2 | XP_011532749.1 | 2 | 912 | |||
| XM_017009139.1 | XP_016864628.1 | 2 | 414 | |||
| XM_017009140.1 | XP_016864629.1 | 2 | 405 | |||
| GenBank mRNA | AK094125.1 | - | 2 | 227 | ||
| AK225523.1 | - | 2 | 237 | |||
| AK314645.1 | - | 2 | 237 | |||
| AL110225.1 | - | 3 | 741 | |||
| BC000283.1 | - | 2 | 220 | 3356428 | ||
| BC007281.1 | - | 2 | 220 | 3343626 | ||
| BC007567.1 | - | 2 | 220 | 3028196 | ||
| BF205663.1 | - | 268 | ||||
| D17530.1 | - | 2 | 221 | |||
| DA245346.1 | - | 2 | 296 | |||
| U00802.1 | - | 2 | 196 | |||

DBN1
Genomic Map