siRNA Details
| Gene Symbol: ZNF100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173531.3 | NP_775802.2 | 2 | 270 | ||
| GenBank mRNA | AK126551.1 | - | 2 | 220 | ||
| AK308798.1 | - | 2 | 191 | |||
| BC035579.1 | - | 2 | 182 | 5576923 | ||
| DA241750.1 | - | 2 | 270 | |||

ZNF100
Genomic Map