siRNA Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139459.1 | NP_001132931.1 | 5 | 1183 | ||
| NM_152609.2 | NP_689822.2 | 5 | 1057 | |||
| XM_005273081.3 | XP_005273138.2 | 5 | 842 | |||
| XM_005273083.4 | XP_005273140.1 | 4 | 760 | |||
| XM_011544110.2 | XP_011542412.1 | 5 | 908 | |||
| XM_011544111.1 | XP_011542413.1 | 5 | 1079 | |||
| XM_011544112.1 | XP_011542414.1 | 4 | 626 | |||
| XM_011544113.1 | XP_011542415.1 | 4 | 751 | |||
| XM_011544114.2 | XP_011542416.1 | 4 | 466 | |||
| XM_017000487.1 | XP_016855976.1 | 6 | 1097 | |||
| XM_017000488.1 | XP_016855977.1 | 5 | 840 | |||
| XM_017000489.1 | XP_016855978.1 | 5 | 907 | |||
| GenBank mRNA | AK056563.1 | - | 5 | 890 | ||
| AK074142.1 | - | 5 | 820 | |||
| AL834246.2 | - | 5 | 891 | |||
| BC010228.2 | - | 5 | 1059 | 4151228 | ||
| BC036200.1 | - | 5 | 950 | |||
| BX647629.1 | - | 5 | 842 | |||

CNST
Genomic Map