siRNA Details
| Gene Symbol: TTC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105669.2 | NP_001099139.2 | 9 | 1519 | ||
| XM_011509253.2 | XP_011507555.1 | 8 | 901 | |||
| XM_011509254.2 | XP_011507556.1 | 8 | 898 | |||
| XM_017000497.1 | XP_016855986.1 | 6 | 523 | |||
| GenBank mRNA | AW628680.1 | - | 3 | 483 | ||

TTC24
Genomic Map