siRNA Details
| Gene Symbol: AP1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030007.1 | NP_001025178.1 | 22 | 2543 | ||
| NM_001128.5 | NP_001119.3 | 21 | 2534 | |||
| GenBank mRNA | AB015317.1 | - | 21 | 2239 | ||
| AK055122.1 | - | 5 | 1154 | |||
| AK128078.1 | - | 24 | 3089 | |||
| AK225906.1 | - | 22 | 2302 | |||
| AK314484.1 | - | 21 | 2539 | |||
| AL110198.1 | - | 5 | 608 | |||
| BC036283.1 | - | 22 | 2508 | 4158263 | ||
| Y12226.1 | - | 22 | 2257 | |||

AP1G1
Genomic Map