siRNA Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207335.2 | NP_997218.2 | 5 | 1706 | ||
| XM_005247167.1 | XP_005247224.1 | 6 | 2170 | |||
| XM_005247168.4 | XP_005247225.1 | 6 | 2248 | |||
| XM_011512490.2 | XP_011510792.1 | 6 | 2170 | |||
| GenBank mRNA | AK128176.1 | - | 5 | 568 | ||
| AK307108.1 | - | 3 | 323 | |||
| BC041365.1 | - | 2 | 65 | |||
| BX537865.1 | - | 5 | 745 | |||

KBTBD12
Genomic Map