siRNA Details
| Gene Symbol: TRIM60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258025.1 | NP_001244954.1 | 4 | 652 | ||
| NM_152620.2 | NP_689833.1 | 3 | 478 | |||
| XM_011531683.2 | XP_011529985.1 | 2 | 472 | |||
| GenBank mRNA | AK093201.1 | - | 3 | 479 | ||
| BC100983.1 | - | 2 | 368 | 40005592 | ||
| BC100984.1 | - | 2 | 368 | 40005593 | ||
| BC100985.1 | - | 2 | 368 | 40005594 | ||
| BC100986.2 | - | 3 | 542 | 40005597 | ||

TRIM60
Genomic Map