siRNA Details
| Gene Symbol: DECR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001359.1 | NP_001350.1 | 8 | 983 | ||
| XM_005250808.4 | XP_005250865.1 | 7 | 671 | |||
| XM_011516880.2 | XP_011515182.1 | 9 | 978 | |||
| XM_017013147.1 | XP_016868636.1 | 10 | 1638 | |||
| XM_017013148.1 | XP_016868637.1 | 10 | 1378 | |||
| GenBank mRNA | AK300069.1 | - | 10 | 1498 | ||
| BC105080.1 | - | 8 | 930 | 8144083 | ||
| BC105082.1 | - | 8 | 930 | 8144085 | ||
| BC143669.1 | - | 6 | 627 | |||
| L26050.1 | - | 8 | 852 | |||
| U49352.1 | - | 8 | 881 | |||

DECR1
Genomic Map