siRNA Details
| Gene Symbol: FREM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168235.1 | NP_001161707.1 | 8 | 6303 | ||
| GenBank mRNA | BE223071.1 | - | 1 | 425 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FREM3| Gene Symbol: FREM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168235.1 | NP_001161707.1 | 8 | 6303 | ||
| GenBank mRNA | BE223071.1 | - | 1 | 425 | ||