siRNA Details
| Gene Symbol: ZNF467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207336.1 | NP_997219.1 | 5 | 1085 | ||
| XM_005249959.4 | XP_005250016.1 | 5 | 1078 | |||
| XM_005249960.4 | XP_005250017.1 | 5 | 1007 | |||
| XM_005249961.4 | XP_005250018.1 | 5 | 1057 | |||
| XM_006715864.3 | XP_006715927.1 | 6 | 1187 | |||
| XM_011515855.2 | XP_011514157.1 | 5 | 2657 | |||
| XM_011515856.2 | XP_011514158.1 | 6 | 2360 | |||
| XM_011515857.2 | XP_011514159.1 | 4 | 2547 | |||
| XM_011515858.1 | XP_011514160.1 | 6 | 1272 | |||
| XM_017011799.1 | XP_016867288.1 | 3 | 2431 | |||
| GenBank mRNA | BC052625.1 | - | 5 | 1085 | 6570263 | |

ZNF467
Genomic Map