siRNA Details
| Gene Symbol: BMPER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133468.4 | NP_597725.1 | 15, 16 | 2248 | ||
| XM_005249633.2 | XP_005249690.1 | 13, 14 | 1658 | |||
| XM_017011800.1 | XP_016867289.1 | 13, 14 | 1808 | |||
| XM_017011801.1 | XP_016867290.1 | 12, 13 | 1478 | |||
| GenBank mRNA | AB075845.1 | - | 11, 12 | 1515 | ||
| AK289963.1 | - | 14, 15 | 1988 | |||
| AY324650.1 | - | 15, 16 | 2247 | |||
| BC060868.1 | - | 14, 15 | 1925 | 30345290 | ||
| BX380517.2 | - | 2, 3 | 215 | |||

BMPER
Genomic Map