siRNA Details
| Gene Symbol: KCNV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133497.3 | NP_598004.1 | 1 | 354 | ||
| GenBank mRNA | AF348983.1 | - | 1 | 144 | ||
| AK302215.1 | - | 1 | 641 | |||
| BC101352.1 | - | 1 | 354 | 40025518 | ||
| BC101353.2 | - | 1 | 354 | 40025524 | ||
| BQ638061.1 | - | 1 | 378 | |||

KCNV2
Genomic Map