siRNA Details
| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010862.2 | NP_001010862.2 | 2 | 910 | ||
| XM_006724577.3 | XP_006724640.1 | 2 | 971 | |||
| XM_006724578.3 | XP_006724641.1 | 2 | 878 | |||
| XM_017029315.1 | XP_016884804.1 | 2 | 1313 | |||
| GenBank mRNA | AK127485.1 | - | 1 | 360 | ||
| AK304047.1 | - | 2 | 847 | |||
| AL832091.1 | - | 2 | 567 | |||
| BC146899.1 | - | 2 | 631 | 9056827 | ||
| BC146906.1 | - | 2 | 631 | 9056834 | ||

SPIN3
Genomic Map