siRNA Details
| Gene Symbol: FAM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164484.1 | NP_001157956.1 | 2 | 887 | ||
| GenBank mRNA | BC029839.1 | - | 2 | 875 | ||
| BC047597.1 | - | 2 | 887 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM170B| Gene Symbol: FAM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164484.1 | NP_001157956.1 | 2 | 887 | ||
| GenBank mRNA | BC029839.1 | - | 2 | 875 | ||
| BC047597.1 | - | 2 | 887 | |||