siRNA Details
| Gene Symbol: SSBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009998.3 | NP_001009998.1 | 2, 3 | 453 | ||
| NM_032627.4 | NP_116016.1 | 2, 3 | 453 | |||
| XM_005259790.3 | XP_005259847.1 | 2, 3 | 494 | |||
| XM_006722665.3 | XP_006722728.1 | 2, 3 | 500 | |||
| XM_006722666.2 | XP_006722729.1 | 2, 3 | 458 | |||
| XM_006722668.2 | XP_006722731.1 | 2, 3 | 458 | |||
| XM_017026431.1 | XP_016881920.1 | 2, 3 | 512 | |||
| XM_017026432.1 | XP_016881921.1 | 2, 3 | 1050 | |||
| XM_017026433.1 | XP_016881922.1 | 2, 3 | 458 | |||
| XM_017026434.1 | XP_016881923.1 | 2, 3 | 507 | |||
| XM_017026435.1 | XP_016881924.1 | 2, 3 | 1049 | |||
| XM_017026436.1 | XP_016881925.1 | 2, 3 | 458 | |||
| XM_017026437.1 | XP_016881926.1 | 2, 3 | 489 | |||
| GenBank mRNA | AK098429.1 | - | 2, 3 | 359 | ||
| BC000274.1 | - | 2, 3 | 360 | 3356283 | ||
| BF207000.1 | - | 2, 3 | 394 | |||
| BQ068310.1 | - | 492 | ||||
| BU838485.1 | - | 2 | 224 | |||
| CX873722.1 | - | 2, 3 | 453 | |||

SSBP4
Genomic Map