siRNA Details
| Gene Symbol: PPP1R18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134870.1 | NP_001128342.1 | 2 | 976 | ||
| NM_133471.3 | NP_597728.1 | 1 | 2622 | |||
| GenBank mRNA | AB073600.1 | - | 2 | 867 | ||
| AB075829.1 | - | 1 | 2040 | |||
| AK097089.1 | - | 1 | 775 | |||
| AK124880.1 | - | 1 | 2622 | |||
| AK125639.1 | - | 2 | 1703 | |||
| AL832742.1 | - | 1 | 1374 | |||
| BC066644.1 | - | 1 | 934 | 5575643 | ||
| BX647622.1 | - | 2 | 876 | |||

PPP1R18
Genomic Map