siRNA Details
| Gene Symbol: ZNF721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133474.3 | NP_597731.2 | 3 | 2093 | ||
| GenBank mRNA | AB075862.1 | - | 1 | 913 | ||
| AK092362.1 | - | 1 | 720 | |||
| AK095558.1 | - | 1 | 254 | |||
| AK127067.1 | - | 1 | 1649 | |||
| AK128499.1 | - | 1 | 1922 | |||
| AL834155.1 | - | 1901 | ||||

ZNF721
Genomic Map