siRNA Details
| Gene Symbol: DIO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000792.6 | NP_000783.2 | 1, 2 | 352 | ||
| NM_001039716.2 | NP_001034805.1 | 1, 2 | 352 | |||
| NR_136693.1 | 2, 3 | 269 | ||||
| GenBank mRNA | AK290780.1 | - | 1, 2 | 352 | ||
| AY560374.1 | - | 1, 2 | 328 | |||
| AY560378.1 | - | 1, 2 | 328 | |||
| AY560380.1 | - | 2, 3 | 204 | |||
| AY560382.1 | - | 2, 3 | 245 | |||
| BC017955.1 | - | 1, 2 | 351 | 4607072 | ||
| BC107170.2 | - | 1, 2 | 351 | 40036824 | ||
| BC107171.2 | - | 1, 2 | 351 | 40036826 | ||
| FJ002243.1 | - | 2, 3 | 267 | |||

DIO1
Genomic Map