siRNA Details
| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004943.1 | NP_004934.1 | 2 | 499 | ||
| GenBank mRNA | BM922103.1 | - | 499 | |||
| L19267.1 | - | 2 | 87 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DMWD| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004943.1 | NP_004934.1 | 2 | 499 | ||
| GenBank mRNA | BM922103.1 | - | 499 | |||
| L19267.1 | - | 2 | 87 | |||