siRNA Details
| Gene Symbol: DXO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005510.3 | NP_005501.2 | 6, 7 | 1382 | ||
| XM_006715005.3 | XP_006715068.1 | 6, 7 | 1461 | |||
| XM_017010329.1 | XP_016865818.1 | 6, 7 | 1165 | |||
| GenBank mRNA | AF059252.1 | - | 6, 7 | 1178 | ||
| BC009344.2 | - | 6, 7 | 1222 | 4130758 | ||
| BC019083.2 | - | 6, 7 | 1394 | 4647012 | ||

DXO
Genomic Map