siRNA Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039589.1 | NP_001034678.1 | 4 | 716 | ||
| NM_001319165.1 | NP_001306094.1 | 5 | 1390 | |||
| NM_001319166.1 | NP_001306095.1 | 5 | 1127 | |||
| NM_001319167.1 | NP_001306096.1 | 5 | 1457 | |||
| NM_001319168.1 | NP_001306097.1 | 5 | 1348 | |||
| NM_001319169.1 | NP_001306098.1 | 4 | 1176 | |||
| NM_001319170.1 | NP_001306099.1 | 5 | 1460 | |||
| NM_001319171.1 | NP_001306100.1 | 5 | 1431 | |||
| NM_001384.4 | NP_001375.2 | 5 | 1400 | |||
| XM_017000502.1 | XP_016855991.1 | 5 | 1402 | |||
| XM_017000503.1 | XP_016855992.1 | 5 | 1336 | |||
| XM_017000504.1 | XP_016855993.1 | 4 | 1641 | |||
| XM_017000505.1 | XP_016855994.1 | 4 | 1573 | |||
| XM_017000506.1 | XP_016855995.1 | 5 | 1444 | |||
| GenBank mRNA | AF053003.1 | - | 5 | 1366 | ||
| AK091303.1 | - | 5 | 1319 | |||
| AK297933.1 | - | 4 | 1149 | |||
| AK315506.1 | - | 5 | 1384 | |||
| BC001389.2 | - | 5 | 1359 | 3051455 | ||
| BC003181.1 | - | 5 | 1377 | 2958202 | ||
| BC016956.1 | - | 5 | 1359 | 3900854 | ||
| BT007431.1 | - | 5 | 1205 | |||

DPH2
Genomic Map