siRNA Details
Gene Symbol: SLC26A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000112.3 | NP_000103.2 | 3 | 1995 | ||
XM_017009191.1 | XP_016864680.1 | 3 | 2003 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC26A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000112.3 | NP_000103.2 | 3 | 1995 | ||
XM_017009191.1 | XP_016864680.1 | 3 | 2003 |